Methylmercaptoacetat 98

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Methyl thioglycolate

L03783.14 L03783.22 L03783.36
L03783.14EA 312 DKK
L03783.14 L03783.22 L03783.36
Methylmercaptoacetat 98
Methylmercaptoacetat

Purity: 98% CAS No.: 2365-48-2 Molecular Formula: C3H6SO2 Formula Weight: 106.14 Flash Point: 56°(132°F) Refractive Index: 1.4660 Odour: Stench Melting Point: -3° Boiling Point: 148-149° Beilstein Reference: 506259 UN No.: 1228 MDL No.: MFCD00004873

Formel: C₃H₆O₂S
Molekylvægt: 106.15 g/mol
Kogepunkt: 148…149 °C
Smeltepunkt: –3 °C
Massefylde: 1.163
Flammepunkt: 56 °C (132 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00004873
CAS-nummer: 2365-48-2
EINECS: 219-121-7
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR