VWR® Traceable®, Full Scale Plus, Digital Thermometers
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
620-2089EA 993 DKK
620-2089 620-2058 620-2064 620-1664
VWR® Traceable®, Full Scale Plus, Digital Thermometers
Termometre Digital termometre
These thermometers, with LCD and magnetic back, enable placement on a metallic surface (incubators, freezers, water baths). They incorporate a flip-open easel for use on a lab bench, and a spring fastener for clipping to any edge.
  • Piercing tip
  • Feature available on 620-1664 and 620-2089 models: Reading updated every second
  • Model 620-2089: High/low alarm, maximum/minimum memories and 'HOLD' function
Stainless steel probe (Ø 3,5 mm) and 3 metre cable.
Certificeringer: Compliant with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Leveringsinformation: More Product Information