Gas distributors and frits for scrubber bottles and columns, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
N1501-16 N1503-32 N1503-40 N1503-36 N1503-28
BOHLN1501-16EA 737 DKK
BOHLN1501-16 BOHLN1503-32 BOHLN1503-40 BOHLN1503-36 BOHLN1503-28
Gas distributors and frits for scrubber bottles and columns, BOLA
Filtre Forkolonnefiltre og fritter
These PTFE gas distributors and frits are suitable for steady sparkling of the flowing gas.
  • Suitable for scrubber bottles and columns with M 8x1 thread and for gas inlet tubes
  • Good chemical resistance
Temperature resistant from –200 to +250 °C
Certificeringer: FDA conform.