Joint clamps, PTFE/steel spring, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
H942-45 H942-32 H942-19 H942-14
BOHLH942-45EA 147 DKK
BOHLH942-45 BOHLH942-32 BOHLH942-19 BOHLH942-14
Joint clamps, PTFE/steel spring, BOLA
Klemmer Klemmer til NS-slib
Reusable clamps with PTFE-encapsulated steel spring core for connecting ground joint parts.
  • Universal chemical resistance since the product is only exposed to PTFE
  • Ideal for use when highly aggressive liquids are involved
  • High recovery even at high temperatures