Tubing, flanged, PTFE, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
F740-24 F740-26 F740-22 F740-04 F740-02 F740-28 F740-06 F740-10 F740-20 F740-08
BOHLF740-24EA 186 DKK
BOHLF740-24 BOHLF740-26 BOHLF740-22 BOHLF740-04 BOHLF740-02 BOHLF740-28 BOHLF740-06 BOHLF740-10 BOHLF740-20 BOHLF740-08
Tubing, flanged, PTFE, BOLA
Slanger Overførselsslange
PTFE, PA. Connection to BOLA distributors for bottles or BOLA chromatography adaptors.
  • Temperature resistance: from 0 to +100 °C
  • Universal chemical resistance
  • Pressure: 30 bar
Flanged PTFE tubing with black tube end fittings UNF ¼" 28 G made of PP and washers made of PA. The tubing is ready for use.