Tubing distributors, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D512-02 D512-08 D512-09 D512-01
BOHLD512-02EA 2640 DKK
BOHLD512-02 BOHLD512-08 BOHLD512-09 BOHLD512-01
Tubing distributors, BOLA
Fittings og stik
Tubing distributors, body made of PTFE with nuts made of glass-fibre reinforced PTFE. One inlet and three or four outlets, bore diameter 6 mmutors made of fluoroplastic with glass-fibre reinforced PTFE nuts.
  • Temperature resistance from −200 to +250 °C
  • Universal chemical resistance
  • Pressure: 5 bar
  • Suitable for vacuum