Cleanroom gloves, nitrile, sterile, BioClean Nitramax™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BNMS-65 BNMS-70 BNMS-85
112-3859EA 27330 DKK
112-3859 112-3860 112-3863
Cleanroom gloves, nitrile, sterile, BioClean Nitramax™
Handsker
These hand-specific nitrile gloves with beaded cuffs provide good resistance against chemicals. They also have excellent ESD properties.
  • Sterile, powder-free, textured finish provides enhanced grip
  • Contain no natural latex proteins, hypoallergenic
  • Extended length provides protection of the entire arm
  • Extended leaching in pure deionised water ensures extremely low levels of chlorides and extractable metals
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
EN 455, EN 374; Cat. III; AQL 1.5
Certificeringer: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Sterilized in accordance with ISO 11137-1:2006.
Suitable for contact with all foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Emballering: One cuffed pair per plastic wallet; one wallet per EasyTear™ PE pouch; 10 pairs per sealed outer PE bag; 10 outer bags per carton liner; one carton liner per carton (100 pairs).