Short ground sockets and cones, finished, DURAN®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
286153606 286531801
SCOT286153606EA 1990 DKK
SCOT286153606 SCOT286531801
Short ground sockets and cones, finished, DURAN®
Adapter Adaptere med NS-slib
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes