Cleanroom gloves, latex, BioClean Legend™
Leverandør: Nitritex
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
BLHN65 BLHN70 BLHN75 BLHN80 BLHN85 BLHN90
NITRBLHN65EA 3740 DKK
NITRBLHN65 NITRBLHN70 NITRBLHN75 NITRBLHN80 NITRBLHN85 NITRBLHN90
Cleanroom gloves, latex, BioClean Legend™
Handsker
These hand-specific cleanroom gloves with beaded cuffs can be used in medical as well as in laboratory environments.
  • Protein level less than 50 µg/g
  • Textured finish for improved grip
  • Latex provides exceptional flexibility and comfort
  • Beaded cuff provides easier donning and stability on the arm
  • Anatomically shaped for enhanced wearer comfort
  • Each size carries a unique bar code on the carton (ITF-14), which allows automatic data entry into stock control systems
  • Cuff marking: Ink Print Size and L or R
  • Glove cleanliness LPC: <2000
  • Standard compliance: PPE, Cat III EN ISO 374 Type B
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
EN 455, EN 374, Cat. III; AQL 1.5
Certificeringer: Manufactured in a facility holding ISO 9001:2015 and (EU)2016/425 Module D certifications.
Conforms to Category 3 - Complex Design Personal Protective Equipment PPE Regulation (EU)2016/425.
Processed in an NEBB certified ISO Class 4 environment.
Complies with the requirements of European Glove Standards EN 420:2003 +A1:2009, EN 374-1:2016, EN 374-5:2016 and EN 421:2010.
Physical properties comply with European Medical Glove Standard EN 455-2:2015.
Suitable for contact with dry, alcoholic and acidic foodstuffs. Regulation (EC)1935/2004 and (EC) 2023/2006.
Emballering: 50 pairs per sealed inner PE bag; one sealed inner PE bag per sealed outer PE bag; four outer PE bags per carton liner; one carton liner per carton (200 pairs).