Percoll™
Leverandør: Cytiva
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
17-0891-01 17-0891-02 17-0891-09
17-0891-01EA 3875 DKK
17-0891-01 17-0891-02 17-0891-09
Percoll™
Celle separations medie
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For Research Use Only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0,25 M sucrose, and for formation of gradients either by the use of convenient gradient mixers or by high speed centrifugation. Colloidal solution of silica coated with polyvinylpyrrolidone (PVP). Liquid/suspension. Colorless to slightly yellow. Certificate of Analysis is available.
Advarsel: Percoll is for Research use only.