PCR plates for Roche® LightCycler® 480, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PCR384LC480WNF PCR-96LC480WBC PCR-96LC480WNF PCR-384-LC480W PCR-96LC480CNF PCR-96-LC480-W
732-1219EA 504 DKK
732-1219 732-1378 732-1379 732-1376 AXYGPCR-96LC480CNF 732-1591
PCR plates for Roche® LightCycler® 480, Axygen®
Mikroplader PCR plader
PP, white. Low profile microplates compatible with many thermal cyclers, but especially suitable for the Roche® LightCycler® 480.