Nephelo culture flasks, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
351456 351454 351486
WHEA351456EA 2020 DKK
WHEA351456 WHEA351454 WHEA351486
Nephelo culture flasks, WHEATON®
Kolber Dyrkningsbeholdere
Nephelo culture flasks, made of borosilicate glass, that conforms to USP Type I and ASTM E 438 Type I, Class A requirements.
  • Scratch and striation free side tube
  • Available with or without cleanout port
  • A 38-430 size top cap and 20-415 size sidearm
  • Cap have white styrene butadiene rubber liner
Designed for use with turbidity meter or nephelometer.