Multiple use earplugs, SmartFit®
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1011239 1012522
DALL1011239EA 831 DKK
DALL1011239 111-0556
Multiple use earplugs, SmartFit®
Ørepropper Genanvendelige ørepropper
These earplugs with Conforming Material Technology™ use body heat to adapt to the shape of the ear canal for a comfortable, personalized fit.
  • Three-flange design delivers superior comfort and a truly individual fit
  • Detachable cord system
  • Metal ring on stem detectable by automated equipment (SmartFit Detectable only)
The detectable version with its bright blue color and metalized features can be detected by visual or automated methods and is therefore ideal for food applications.
SNR: 25 dB
Certificeringer: EN 352-2
Leveringsinformation: More Product Information