Screw thread connectors, Wheaton Connection®, WHEATON®
Leverandør: DWK Life Sciences
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
125486 125478 125480 125488 125492 125494 125496 125497 125499 125500 125504 125482
WHEA125486EA 2600 DKK
WHEA125486 WHEA125478 WHEA125480 WHEA125488 WHEA125492 WHEA125494 WHEA125496 WHEA125497 WHEA125499 WHEA125500 WHEA125504 WHEA125482
Screw thread connectors, Wheaton Connection®, WHEATON®
Fittings og stik
Connectors for threaded pipes, with white polypropylene or black phenolic caps. Connectors can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere.
  • Inert contact surface
  • Easy to assemble
  • Screw cap configuration provides a grease-free, vacuum-tight seal
  • safely joins two exterior threaded glass components without hooks, springs or clamps
  • Working temperature up to 180 °C