Reflectoquant® teststrimler, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.16892.0001 1.16899.0001 1.16977.0001
1.16892.0001EA 916 DKK
1.16892.0001 1.16899.0001 1.16977.0001
Reflectoquant® teststrimler, Supelco®
Miljø test kit
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly