Goggles, Honeywell LG
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1005507 1005509 1005504 1008112
111-1231EA 36.8 DKK
111-1231 111-1232 DALL1005504 DALL1008112
Goggles, Honeywell LG
Beskyttelsesbriller
The impact resistant polycarbonate lens offers an excellent protection against flying particles, coarse dusts, liquid droplets, mists and sprays.
  • Lightweight design and large field of vision
  • Soft seal and easily adjustable headband: Fits a wide variety of workers
  • With or without ventilation
The acetate, anti-fog lens and neoprene headband offers superior chemical resistance allowing the use in a broad range of chemical industries.
Weight: 75 to 79 g
Certificeringer: EN 166