Goggles, DuraMaxx™
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1017737 1017750
111-1433EA 0 DKK
111-1433 111-1435
Goggles, DuraMaxx™
Beskyttelsesbriller
These low profile goggles with toric lens provide superior optics and unobstructed vision, while the soft, elastomer bodies flex and conform to the face for a comfortable, gap-free fit.
  • PP/TPE frame and large adjustable headband
  • Indirect venting system minimises fogging to keep lenses clear
  • Dura-streme™ lens coating provides anti-scratch coating on the outside and anti-fog coating on the inside
The OTG goggles fit over most prescription spectacles comfortably. Clear lens model is adapted for indoor use, grey model for outdoor use.
Weight: 100 g
Certificeringer: EN 166