VWR®, pulverpakker til bestemmelse af nitrit, reagenser til fotometre
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
85613.620EA 332 DKK
85613.620 85613.680 85614.620
VWR®, pulverpakker til bestemmelse af nitrit, reagenser til fotometre
Miljø test kit
VWR® powder pack reagents are designed to be used on laboratory and portable photometers with 16 mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminium foil packs, are unaffected by ambient conditions and contain pre-measured powder reagents for accuracy, convenience and great solubility.
  • Accurate
  • Fast and convenient by easier dissolution than tablets and long shelf life
  • No calibration necessary
  • Beveled edge prevents reagent from getting stuck in corners