Overalls, Tyvek® 500 Labo, model CHF7
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TYCHF7SWH2X002 TYCHF7SWHSM002 TYCHF7SWHMD002 TYCHF7SWHLG002 TYCHF7SWHXL002 TYCHF7SWH3X002
113-1551EA 3440 DKK
113-1551 113-1547 113-1548 113-1549 113-1550 113-1552
Overalls, Tyvek® 500 Labo, model CHF7
Engangsbeklædning
These garments are made of Tyvek®, a fabric composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Tyvek® is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres (down to 1 µ in size).
  • Ultra-low-linting fabric
  • Tyvek® zipper and zipper flap for enhanced protection
  • 3-piece hood and gusset for improved fit
  • Elasticated face, wrists and ankles as well as glued-in waist elastic
  • Attached shoe covers consisting of garment material with friction-coated sole to provide added slip-resistance
  • Stitched internal seams to help reduce contamination from the inside to the outside of the garment
  • Antistatic treatment (EN 1149-5) - on both sides
  • Robust yet lightweight (<250 g per garment)
Typical applications include the pharmaceutical industry, laboratories, cosmetics, optical and electronics.
Colour: White.
Weight: 41,5 g/m².
Certificeringer: Type 5/6, EN 1149-5, EN 1073-2; Cat. III.; (EU) 2016/425.
Emballering: 25 per box, individually packed.