Overalls, Tyvek® 400 Dual, model CHF5a
Leverandør: DuPont
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TDCHF5SWH3X010 TDCHF5SWHLG010 TDCHF5SWHMD010 TDCHF5SWHSM010 TDCHF5SWHXL010 TDCHF5SWH2X010
113-9710EA 12440 DKK
113-9710 113-9707 113-9706 113-9705 113-9708 113-9709
Overalls, Tyvek® 400 Dual, model CHF5a
Engangsbeklædning
The Tyvek® 400 Dual garment combines Tyvek® on the front with a non woven SMS panel on the back. The Tyvek® front is permeable to both air and water vapour, yet repels water-based liquids and aerosols. It offers an excellent barrier against fine particles and fibres down to 1 micron in size.
  • Tyvek® front is ultra-low-linting
  • Antistatic treatment (EN 1149-5) - on inside
  • Tyvek® zipper and zipper flap for enhanced protection
  • 3-piece hood and gusset for optimal fit
  • Elasticated face, wrists, ankles and waist (stitched-in)
  • Stitched external seams for enhanced protection against penetration from the outside to the inside of the garment
The large, breathable back panel, made of non woven SMS, offers lower protection against particles (down to 3 microns in size) and light water-based splashes, yet high levels of comfort.
The overall is designed for specific applications that demand comfort while helping to provide protection from frontal exposure during brick ceramic firing, foundries and smelting operations, paint spraying or any work involving composite materials, glass manufacturing, and utilities.
Weight: 41,5/60 g/m²
Certificeringer: Type 5/6, EN 1149-5, EN 1073-2, Cat. III.; (EU) 2016/425
Emballering: 100 per box, individually packed.