HPLC columns, Chromolith® HighResolution RP-18e
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
1.52020.0001 1.52021.0001 1.52022.0001 1.52025.0001 1.52019.0001 1.52023.0001 1.52064.0001 1.52320.0001 1.52321.0001 1.52322.0001 1.52325.0001
1.52020.0001EA 4580 DKK
1.52020.0001 1.52021.0001 1.52022.0001 1.52025.0001 1.52019.0001 1.52023.0001 1.52064.0001 1.52320.0001 1.52321.0001 1.52322.0001 1.52325.0001
HPLC columns, Chromolith® HighResolution RP-18e
Kromatografikolonner
Pore volume: 2,9 ml/g.
  • High efficiencies at lower back pressure
  • Improved peak symmetry
  • Can be used for standard HPLC/UHPLC/LC-MS instruments
  • Back pressure is two times lower compared to particulate-packed columns
  • 30% longer column lifetime compared to particulate-packed columns
Macropore size: 1,15 µm.
Mesopore size: 15 nm.
Surface area: 250 m²/g.
Pore volume: 2,9 ml/g