2,2-Bis(hydroxymethyl)propionsyre for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid
M-Clarity™ quality level = MQ200
8.14996.0250
8.14996.1000
8.14996.0250EA
597
DKK
8.14996.0250
8.14996.1000
2,2-Bis(hydroxymethyl)propionsyre for synthesis, Sigma-Aldrich®
2,2-Bis(hydroxymethyl)propionsyre
Formel:
C₅H₁₀O₄ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004199 CAS-nummer: 4767-03-7 EINECS: 225-306-3 |
Specifikationer Testresultater
Solubility | (20 °C) soluble |
Melting Point | 188 - 191 °C |
Molar Mass | 134.13 g/mol |
Flash Point |
|
pH | 2.6 (50 g/l, H2O, 20 °C) |
Ignition Point |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...