Diisoamylether (mix af isomere) 96%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A11231.AC A11231.AE A11231.AP
A11231.ACEA 340 DKK
A11231.AC A11231.AE A11231.AP
Diisoamylether (mix af isomere) 96%
Diisoamylether (mix af isomere)

Purity: 96% CAS No.: 544-01-4 Molecular Formula: [(CH3)2CHCH2CH2]2O Formula Weight: 158.29 Flash Point: 46°(115°F) Refractive Index: 1.407 Boiling Point: 172-174° Beilstein Reference: 1698014 Merck Reference: 14,5118 UN No.: 3271 MDL No.: MFCD00008947

Formel: C₁₀H₂₂O
Kogepunkt: 172…174 °C
Massefylde: 0,78
Flammepunkt: 46 °C (115 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00008947
CAS-nummer: 544-01-4
EINECS: 208-857-4
UN nummer: 3271
ADR: 3,III
Merck Index: 14,5118

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR