Iod(I)chlorid 1 M dichloromethane
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Iodochloride
370741000 370748000
ACRO370741000EA 762 DKK
ACRO370741000 ACRO370748000
Iod(I)chlorid 1 M dichloromethane
Iodine monochloride
Formel: ClI
Molekylvægt: 162.36 g/mol
Kogepunkt: 97.3
Smeltepunkt: 26
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011354
CAS-nummer: 7790-99-0
EINECS: 232-236-7
UN nummer: 2922
ADR: 8 II
Merck Index: 13,05039
Bestil nu
Specifikationer Testresultater
Appearance Orange to red to brown Solution
Titration with Na2S2O3 0.95 to 1.05 M