Bolts and clamp rings
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C410-08 C410-04 C410-06 C410-02
BOHLC410-08EA 135 DKK
BOHLC410-08 BOHLC410-04 BOHLC410-06 BOHLC410-02
Bolts and clamp rings
Klemmer Universalklemmer
Bolts and clamps made entirely of PTFE, which are mainly used for moon-shaped stirrer blades with double-sided groove.
  • Temperature resistance: −200 to 250 °C
  • Universal chemical resistance