2-Chlor-3-nitropyridin ≥99%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

A12161.06 A12161.14 A12161.22
A12161.06EA 370 DKK
A12161.06 A12161.14 A12161.22
2-Chlor-3-nitropyridin ≥99%
2-Chloro-3-nitropyridine

Purity: 99% CAS No.: 5470-18-8 Molecular Formula: C5H3ClN2O2 Formula Weight: 158.54 Flash Point: 185°(365°F) Melting Point: 99-103° Beilstein Reference: 124482 MDL No.: MFCD00006232

Formel: C₅H₃ClN₂O₂
Kogepunkt: 260 °C (1013 hPa)
Smeltepunkt: 99…103 °C
Flammepunkt: 185 °C (365 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006232
CAS-nummer: 5470-18-8
EINECS: 226-799-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR