2-Chlor-3-nitropyridin 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

109730250 109731000
ACRO109730250EA 717 DKK
ACRO109730250 ACRO109731000
2-Chlor-3-nitropyridin 99+%
2-Chloro-3-nitropyridine
Formel: C₅H₃ClN₂O₂
Kogepunkt: 260 °C (1013 hPa)
Smeltepunkt: 100…103 °C
Flammepunkt: 185 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006232
CAS-nummer: 5470-18-8
EINECS: 226-799-8

Bestil nu

Specifikationer Testresultater

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 103 °C
HPLC ≥99.0 %
Sulfate (SO4) ≤0.002 %
Ammonium (NH4) ≤0.002 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR