Overhead stirrer shafts, with blade, BOLA
Leverandør: Bohlender
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
C381-04 C381-08 C381-10 C381-06 C381-12
BOHLC381-04EA 2800 DKK
BOHLC381-04 BOHLC381-08 BOHLC381-10 BOHLC381-06 BOHLC381-12
Overhead stirrer shafts, with blade, BOLA
Omrørere Omrøreaksler
Stirrer shafts made of PTFE-jacketed stainless steel with the blade made completely of PTFE.
  • Chemical resistant
  • Temperature resistance from –200 to +250 °C
  • Creates tangential flow with little turbulence