Tetrahydrofuran

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

7344.1 7344.2
ROTH7344.1EA 0 DKK
ROTH7344.1 ROTH7344.2
Tetrahydrofuran
Tetrahydrofuran
Formel: C₄H₈O
Kogepunkt: 66 °C (1013 hPa)
Massefylde: 0,888 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00005356
CAS-nummer: 109-99-9
EINECS: 203-726-8
UN nummer: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR