Industrial gloves, Polytril™ Black
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2232231/10 2232231/7 2232231/8 2232231/9
DALL2232231/10EA 0 DKK
DALL2232231/10 DALL2232231/7 DALL2232231/8 DALL2232231/9
Industrial gloves, Polytril™ Black
Handsker
Black polyamide knitted glove with black oil proof nitrile coating on palm and fingertips and elastic wrist.
  • Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back improves the glove's breathability
  • Nitrile coating provides outstanding abrasion resistance and impermeability to oils
  • Filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
The glove is suitable for the handling of fine products in a greasy, damp or dirty environment and mechanical handling in the automotive industry.
Certificeringer: EN 388:2016 (3121X); Cat. II