Industrial gloves, Polytril™
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2232230/9 2232230/10 2232230/7 2232230/8
112-1523EA 33.2 DKK
112-1523 112-1517 112-1518 112-1519
Industrial gloves, Polytril™
Handsker
White polyamide knitted glove with grey oilproof nitrile coating on palm and fingertips and elastic knitted wrist.
  • Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
  • The ventilated back improves the glove's breathability
  • Knitted and elasticated wrist gives the glove good support
  • Nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
Suitable for handling of fine objects in a greasy, damp or soiled atmosphere and mechanical handling in the automotive industry.
Certificeringer: EN 388:2016 (4121X); Cat. II.