General purpose gloves, Perfect Poly® grey
Leverandør: HONEYWELL SAFETY
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2400250/10 2400250/11 2400250/8 2400250/9
DALL2400250/10EA 34.6 DKK
DALL2400250/10 DALL2400250/11 DALL2400250/8 DALL2400250/9
General purpose gloves, Perfect Poly® grey
Handsker
Grey polyamide knitted glove with grey polyurethane coating on the fingertips that makes the glove suitable for precision work.
  • Polyamide with its polyurethane coating provide excellent abrasion and tear resistance
  • Ventilated back and palm optimise the glove's breathability
  • Non pilling, the polyamide is suitable for precision work
  • Its dark colour makes it more soilproof, thus ensuring a longer life
Ideal for handling fine products in a dry or slightly greasy or soiled environment.
Certificeringer: EN 388:2016 (4121X); Cat. II