2,7-Dimethylnaphthalen ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
B21704.01
B21704.03
B21704.01EA
879
DKK
B21704.01
B21704.03
2,7-Dimethylnaphthalen ≥99%
2,7-Dimethylnaphthalen
Purity: 99%
CAS No.: 582-16-1
Molecular Formula: C12H12
Formula Weight: 156.23
Melting Point: 96-98°
Boiling Point: 262-264°
MDL No.: MFCD00004121
Formel:
C₁₂H₁₂ Kogepunkt: 262…264 °C Smeltepunkt: 96…98 °C Massefylde: 1,003 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004121 CAS-nummer: 582-16-1 EINECS: 209-477-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...