2-(Brommethyl)naphthalen 96%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
B24568.06
B24568.14
B24568.22
B24568.06EA
513
DKK
B24568.06
B24568.14
B24568.22
2-(Brommethyl)naphthalen 96%
2-(Brommethyl)naphthalen
Purity: 96%
CAS No.: 939-26-4
Molecular Formula: C11H9Br
Formula Weight: 221.10
Storage Temperature: 2°C to 8°C
Flash Point:
Formel:
C₁₁H₉Br Molekylvægt: 221.1 g/mol Kogepunkt: 213 °C (100 mmHg) Smeltepunkt: 51…55 °C Flammepunkt: >110 °C (230 °F) Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00004123 CAS-nummer: 939-26-4 EINECS: 213-359-5 UN nummer: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...