Petri dishes, Sterilin®
Leverandør: STERILIN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
101VR20 101RT/C 101R20 101/IRR 101V/IRR 101RT/IRR 101VR20/C
391-2016EA 1710 DKK
391-2016 391-0445 391-2015 391-0539 391-0541 391-0540 SLIN101VR20/C
Petri dishes, Sterilin®
Dyrkningsplader Petriskåle
Non cytotoxic, virgin PS. Standard 90 mm Petri dishes are used by microbiologists to culture microorganisms on solid media. Triple vented dishes aid gaseous exchange and are ideal for short term work. Single vented dishes limit gaseous exchange, minimise evaporation and dehydration, and are ideally suited for long term work. Non vented dishes are most appropriate for anaerobic and long term work.
  • Mirror finished moulds ensure high optical clarity
  • Available either aseptically manufactured or terminally sterilised by gamma irradiation
  • Ideal for use in automatic plate pourers
  • Available in single, triple or non vented formats
Produced and tested in accordance with EN ISO 24998:2008 Petri dish standard, which includes stringent dimentional controls.
Dimensions shown are for outer Ø base and outer height (mm).
Culture microorganisms on solid media using Thermo Scientific™ Sterilin™ standard 90 mm Petri dishes. These shallow dishes maximise incubator space and feature a mirror finish that ensures high optical quality. Not available in America.