Zirconiumnitrid ≥99.5% (metals basis excluding Hf) (max. 3% Hf)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
12138.14
12138.22
12138.14EA
2050
DKK
12138.14
12138.22
Zirconiumnitrid ≥99.5% (metals basis excluding Hf) (max. 3% Hf)
Zirconiumnitrid
Purity: 99.5%
CAS No.: 25658-42-8
Molecular Formula: NZr
Formula Weight: 105.23
Physical Form: -325 Mesh Powder
Density: 7.09
Appearance: Yellow-brown
Odour: Odourless
Melting Point: 2980°
UN No.: 3178
MDL No.: MFCD00049642
Formel:
NZr Smeltepunkt: 2980 °C Massefylde: 7,09 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00049642 CAS-nummer: 25658-42-8 EINECS: 247-166-2 UN nummer: 3178 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...