Tellurdioxid 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

222590500 222592500
ACRO222590500EA 897 DKK
ACRO222590500 ACRO222592500
Tellurdioxid 99+%
Tellurdioxid
Formel: O₂Te
Kogepunkt: 1245 °C (1013 hPa)
Smeltepunkt: 733 °C
Massefylde: 5,6 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011263
CAS-nummer: 7446-07-3
EINECS: 231-193-1
UN nummer: 3077
Merck Index: 14,09123

Bestil nu

Specifikationer Testresultater

Appearance White to light yellow Powder
Assay ≥99.0 % (TeO2)
Copper (Cu) ≤50 ppm
Iron (Fe) ≤50 ppm
Lead (Pb) ≤50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR