Bismuth(III)tellurid ≥99.999% (metals basis) for vacuum deposition
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Purity: 99.999%
CAS No.: 1304-82-1
Molecular Formula: Bi2Te3
Formula Weight: 800.76
Physical Form: 6mm & Down Pieces
Sensitivity: Moisture Sensitive
Appearance: Grey
Odour: Odourless
Melting Point: 573°
UN No.: 3284
MDL No.: MFCD00014201
Formel:
Bi₂Te₃ Smeltepunkt: 573 °C Massefylde: 7,69 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00014201 CAS-nummer: 1304-82-1 EINECS: 215-135-2 UN nummer: 3284 ADR: 6.1,III Merck Index: 12,01331 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...