Tintetrachlorid, anhydrous ≥98% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Stannic chloride, Tin tetrachloride

11571.36 11571.57
11571.36EA 1310 DKK
11571.36 11571.57
Tintetrachlorid, anhydrous ≥98% (metals basis)
Tintetrachlorid

Purity: 98% CAS No.: 7646-78-8 Molecular Formula: Cl4Sn Formula Weight: 260.50 Physical Form: Liquid Density: 2.26 Refractive Index: 1.512 Sensitivity: Moisture Sensitive Appearance: Colourless Melting Point: -33° Boiling Point: 114° Solubility: Soluble in alcohol, CCl4, benzene, toluene, and acetone. Soluble in water with much heat evolution Merck Reference: 14,8774 UN No.: 1827 MDL No.: MFCD00011242

Formel: Cl₄Sn
Kogepunkt: 114 °C
Smeltepunkt: –33 °C
Massefylde: 2,26
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011242
CAS-nummer: 7646-78-8
EINECS: 231-588-9
UN nummer: 1827
ADR: 8,II
Merck Index: 14,8774

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR