Borsyre

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

15165.01 15165.02
SERA15165.01EA 305.8 DKK
SERA15165.01 SERA15165.02
Borsyre
Borsyre
Formel: B(OH)₃
Molekylvægt: 61.83 g/mol
Kogepunkt: 185
Smeltepunkt: 160
Massefylde: 1.44 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011337
CAS-nummer: 10043-35-3
EINECS: 233-139-2
REACH: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR