Di-sec-butylamin ≥99.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D1604-25ML
D1604-500ML
TCIAD1604-25MLEA
132
DKK
TCIAD1604-25ML
TCIAD1604-500ML
Di-sec-butylamin ≥99.0% (by GC)
Di-sec-butylamin
Formel:
C₈H₁₉N Kogepunkt: 132°C Flammepunkt: 24°C |
MDL Number:
MFCD00009326 CAS-nummer: 626-23-3 EINECS: 210-937-9 UN nummer: 2734 ADR: 8,II |
|
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...