Thallium(I)chlorid, ultra dry ≥99.999% (metals basis)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Thallium chloride
Ampuled under argon
35721.06
35721.14
35721.06EA
2380
DKK
35721.06
35721.14
Thallium(I)chlorid, ultra dry ≥99.999% (metals basis)
Thallium(I) chloride
Purity: 99.999%
CAS No.: 7791-12-0
Molecular Formula: TlCl
Formula Weight: 239.82
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 7.0
Refractive Index: 2.247
Sensitivity: Air Sensitive
Melting Point: 430°
Merck Reference: 14,9260
UN No.: 1707
MDL No.: MFCD00011274
Formel:
ClTl Molekylvægt: 239.84 g/mol Kogepunkt: 720 Smeltepunkt: 430 Massefylde: 7.0 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00011274 CAS-nummer: 7791-12-0 EINECS: 232-241-4 UN nummer: 1707 ADR: 6.1 II Merck Index: 14,9260 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...