Crotonaldehyd mixture of isomers ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 2-Butenal, mixture of cis and trans, Crotonaldehyde, mixture of cis and trans
M-Clarity™ quality level = MQ200
8.02667.0100 8.02667.1000
8.02667.0100EA 416 DKK
8.02667.0100 8.02667.1000
Crotonaldehyd mixture of isomers ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
Crotonaldehyd
Formel: C₄H₆O
Molekylvægt: 70.09 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00007003
CAS-nummer: 4170-30-3
EINECS: 224-030-0
UN nummer: 1143
ADR: 6.1 (3) I