Methyl5-methylnicotinat 94

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 5-methylnicotinate

L16371.03 L16371.06
L16371.03EA 602 DKK
L16371.03 L16371.06
Methyl5-methylnicotinat 94
Methyl5-methylnicotinat

Purity: 94% CAS No.: 29681-45-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Melting Point: 42-46° Boiling Point: 90°/3mm MDL No.: MFCD01075739

Formel: C₈H₉NO₂
Molekylvægt: 151.16 g/mol
Kogepunkt: 90 °C (3 mmHg)
Smeltepunkt: 42…46 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01075739
CAS-nummer: 29681-45-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR