Triethylene glycol dimethyl ether ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
2,5,8,11-Tetraoxadodecane
L04128.22
L04128.36
L04128.0E
L04128.22EA
326
DKK
L04128.22
L04128.36
L04128.0E
Triethylene glycol dimethyl ether ≥99%
Triethylene glycol dimethyl ether
Purity: 99%
CAS No.: 112-49-2
Molecular Formula: C8H18O4
Formula Weight: 178.23
Flash Point: 113°(235°F)
Refractive Index: 1.4230
Melting Point: -45°
Boiling Point: 215-216°
Beilstein Reference: 1700630
Merck Reference: 14,9692
MDL No.: MFCD00008504
Formel:
C₈H₁₈O₄ Molekylvægt: 178.23 g/mol Kogepunkt: 215…216 °C Smeltepunkt: –45 °C Massefylde: 0.986 Flammepunkt: 113 °C (235 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00008504 CAS-nummer: 112-49-2 EINECS: 203-977-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...