(Triisopropylsilyl)acetylen ≥97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethynyltriisopropylsilane

H53405.06 H53405.14
H53405.06EA 1260 DKK
H53405.06 H53405.14
(Triisopropylsilyl)acetylen ≥97%
(Triisopropylsilyl)acetylen

Purity: 97% CAS No.: 89343-06-6 Molecular Formula: C11H22Si Formula Weight: 182.38 Flash Point: 56°(133°F) Refractive Index: 1.4527 Boiling Point: 50-52°/0.6mm Beilstein Reference: 3536241 UN No.: 1993 MDL No.: MFCD00075452

Formel: C₁₁H₂₂Si
Molekylvægt: 182.38 g/mol
Kogepunkt: 50…52 °C (0.6 mmHg)
Massefylde: 0.813
Flammepunkt: 56 °C (133 °F)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00075452
CAS-nummer: 89343-06-6
ADR: 3 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR