Dibenzylsuccinat ≥98%
Leverandør: Thermo Fisher ScientificSamlet vurdering
Gennemsntilig vurdering
Purity: 98%
CAS No.: 103-43-5
Molecular Formula: C18H18O4
Formula Weight: 298.34
Refractive Index: 1.5960
Melting Point: 42-47°
Boiling Point: 245°/15mm
Beilstein Reference: 1916056
MDL No.: MFCD00022034
Formel:
C₁₈H₁₈O₄ Kogepunkt: 245 °C (15 mmHg) Smeltepunkt: 42…47 °C Massefylde: 1,256 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00022034 CAS-nummer: 103-43-5 EINECS: 203-110-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...