Methyl6-methoxynicotinat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Methyl 6-methoxynicotinate, 6-Methoxynicotinic acid methylester, Methyl 2-methoxy-5-pyridinecarboxylate

B20808.18 B20808.09 B20808.30
B20808.18EA 2670 DKK
B20808.18 B20808.09 B20808.30
Methyl6-methoxynicotinat ≥98%
Methyl6-methoxynicotinat

Purity: 98% CAS No.: 26218-80-4 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: ca 48-53° MDL No.: MFCD00674130

Formel: C₈H₉NO₃
Molekylvægt: 167.16 g/mol
Smeltepunkt: ∼ 48…53 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00674130
CAS-nummer: 26218-80-4

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR