Acetamidin, HCl ≥97%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
A16091.36
A16091.14
A16091.22
A16091.36EA
1900
DKK
A16091.36
A16091.14
A16091.22
Acetamidin, HCl ≥97%
Acetamidin, HCl
Purity: 97%
CAS No.: 124-42-5
Molecular Formula: C2H6N2.HCl
Formula Weight: 94.54
Melting Point: 168-175°
Beilstein Reference: 3591762
Merck Reference: 14,44
MDL No.: MFCD00013016
Formel:
C₂H₇ClN₂ Molekylvægt: 94.54 g/mol Smeltepunkt: 168…175 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00013016 CAS-nummer: 124-42-5 EINECS: 204-700-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...