Indiumarsenid ≥99.9999% (metals basis)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

12932.04 12932.09
12932.04EA 1800 DKK
12932.04 12932.09
Indiumarsenid ≥99.9999% (metals basis)
Indiumarsenid

Purity: 99.9999% CAS No.: 1303-11-3 Molecular Formula: InAs Formula Weight: 189.74 Physical Form: 1.5-9.5mm Polycrystalline Pieces Odour: Odourless Merck Reference: 14,4949 UN No.: 1557 MDL No.: MFCD00016144

Formel: AsIn
Molekylvægt: 189.74 g/mol
Smeltepunkt: 936
Massefylde: 5.67 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00016144
CAS-nummer: 1303-11-3
EINECS: 215-115-3
UN nummer: 1557
ADR: 6.1 III
Merck Index: 14,4949

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR